2-Chloro-1-(4-fluorobenzyl)benzimidazole ≥97%

Supplier: BeanTown Chemical

Synonyms: 2-Chloro-1-(4-fluorobenzyl)-1H-benzo[d]imidazole

141220-1G 141220-5G
BT141220-1GEA 27 USD
BT141220-1G BT141220-5G
2-Chloro-1-(4-fluorobenzyl)benzimidazole ≥97%
2-Chloro-1-(4-fluorobenzyl)benzimidazole

CAS: 84946-20-3; MDL No: MFCD00800240 Solid; Molecular Formula: C14H10ClFN2; MW: 260.69 Melting Point: 83-87°


  • Solid

Formula: C₁₄H₁₀ClFN₂
MW: 260.70 g/mol
Melting Pt: 83…87 °C
MDL Number: MFCD00800240
CAS Number: 84946-20-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR