1,8-Octanediol ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,8-Dihydroxy octane
141185-100G
141185-25G
141185-500G
BT141185-100GEA
105
USD
BT141185-100G
BT141185-25G
BT141185-500G
1,8-Octanediol ≥98%
1,8-Octanediol
CAS: 629-41-4; EC No: 211-090-8; MDL No: MFCD00002989
Powder; Linear Formula: HO(CH2)8OH; MW: 146.23
Melting Point: 57-61°; Boiling Point: 172°/20 mmHg; Flash point: 120°C (248°F)
- Powder
Formula:
C₈H₁₈O₂ MW: 146.23 g/mol Boiling Pt: 172 °C/20 mmHg Melting Pt: 57…61 °C Flash Pt: 120 °C (248 °F) |
MDL Number:
MFCD00002989 CAS Number: 629-41-4 EINECS: 211-090-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...