4-Bromo-2-nitrobenzoic acid ≥97%
Supplier: BeanTown Chemical
141175-1G
141175-5G
BT141175-1GEA
39
USD
BT141175-1G
BT141175-5G
4-Bromo-2-nitrobenzoic acid ≥97%
4-Bromo-2-nitrobenzoic acid
CAS: 99277-71-1; MDL No: MFCD01013599
Solid; Molecular Formula: C7H4BrNO4; MW: 246.01
Melting Point: 165-169°
- Solid
Formula:
C₇H₄BrNO₄ Melting Pt: 165…169 °C |
MDL Number:
MFCD01013599 CAS Number: 99277-71-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...