Tannic acid

Supplier: BeanTown Chemical

Synonyms: Gallotannin, Tannin

141145-1KG 141145-250G
BT141145-1KGEA 150.31 USD
BT141145-1KG BT141145-250G
Tannic acid
Tannic acid

CAS: 1401-55-4; EC No: 215-753-2; MDL No: MFCD00066397; RTECS: WW5075000 Solid; Molecular Formula: C76H52O46; MW: 1701.20 Melting Point: 218°; Flash point: 199°C (390°F)


  • RTECS: WW5075000
  • Solid

Formula: C₇₆H₅₂O₄₆
MW: 1701.2 g/mol
Melting Pt: 218 °C
Flash Pt: 199 °C (390 °F)
MDL Number: MFCD00066397
CAS Number: 1401-55-4
EINECS: 215-753-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR