Methyl-3-oxotetrahydrothiophene-2-carboxylate ≥97%

Supplier: BeanTown Chemical

Synonyms: Methyl 3-oxotetrahydrothiophene-2-carboxylate

141135-1G 141135-250MG 141135-5G
BT141135-1GEA 200 USD
BT141135-1G BT141135-250MG BT141135-5G
Methyl-3-oxotetrahydrothiophene-2-carboxylate ≥97%
Methyl-3-oxotetrahydrothiophene-2-carboxylate

CAS: 2689-69-2; EC No: 220-257-4; MDL No: MFCD00156156 Solid; Molecular Formula: C6H8O3S; MW: 160.19 Melting Point: 38°; Boiling Point: 232-254°/0.1 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.281; Refractive Index: 1.511


  • Refractive index: 1.511
  • Solid

Formula: C₆H₈O₃S
MW: 160.187 g/mol
Boiling Pt: 232…254 °C/0.1 mmHg
Melting Pt: 38 °C
Density: 1.281 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00156156
CAS Number: 2689-69-2
EINECS: 220-257-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR