7-Bromoisatin ≥97%

Supplier: BeanTown Chemical

Synonyms: 7-Bromo-1H-indole-2,3-dione, 7-Bromoindoline-2,3-dione

141105-1G 141105-5G
BT141105-1GEA 79 USD
BT141105-1G BT141105-5G
7-Bromoisatin ≥97%
7-Bromoisatin

CAS: 20780-74-9; MDL No: MFCD00774354 Solid; Molecular Formula: C8H4BrNO2; MW: 226.03 Melting Point: 191-198°


  • Solid

Formula: C₈H₄BrNO₂
MW: 226.03 g/mol
Melting Pt: 191…198 °C
MDL Number: MFCD00774354
CAS Number: 20780-74-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR