1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,2,3,4-Tetrahydro-1-naphthoic acid

141015-1G 141015-250MG
BT141015-1GEA 239 USD
BT141015-1G BT141015-250MG
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid ≥98%
1,2,3,4-Tetrahydronaphthalene-1-carboxylic acid

CAS: 1914-65-4; MDL No: MFCD02179127 Solid; Molecular Formula: C11H12O2 ; MW: 176.21


  • Solid

Formula: C₁₁H₁₂O₂
MW: 176.22 g/mol
MDL Number: MFCD02179127
CAS Number: 1914-65-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR