Dimethyl-2,6-pyridinedicarboxylate ≥98%

Supplier: BeanTown Chemical

Synonyms: Dimethyl pyridine-2,6-dicarboxylate

141010-10G 141010-50G
BT141010-10GEA 65 USD
BT141010-10G BT141010-50G
Dimethyl-2,6-pyridinedicarboxylate ≥98%
Dimethyl 2,6-Pyridinedicarboxylate

CAS: 5453-67-8; EC No: 226-697-3; MDL No: MFCD00134493 Solid; Molecular Formula: C9H9NO4; MW: 195.17 Melting Point: 121-125°


  • Solid

Formula: C₉H₉NO₄
Melting Pt: 121…125 °C
MDL Number: MFCD00134493
CAS Number: 5453-67-8
EINECS: 226-697-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR