Dithizone ≥85% ACS

Supplier: BeanTown Chemical

140990-25G 140990-5G
BT140990-25GEA 146.22 USD
BT140990-25G BT140990-5G
Dithizone ≥85% ACS
Dithizone

CAS: 60-10-6; EC No: 200-454-1; MDL No: MFCD00003025; RTECS: LQ9450000 Crystalline; Linear Formula: C6H5NHNHCSN=NC6H5; Molecular Formula: C13H12N4S; MW: 256.33 Melting Point: 168° (decomposes)


  • RTECS: LQ9450000
  • Crystalline

Formula: C₆H₅NHNHCSN=NC₆H₅
Melting Pt: 168 °C (dec.)
MDL Number: MFCD00003025
CAS Number: 60-10-6
EINECS: 200-454-1

Order Now

Specification Test Results

Assay 85.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR