3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
Phenazopyridine hydrochloride, 3-Phenylazo-2,6-diaminopyridine hydrochloride
140960-1G
140960-25G
140960-5G
BT140960-1GEA
32
USD
BT140960-1G
BT140960-25G
BT140960-5G
3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride ≥98%
3-(Phenyldiazenyl)pyridine-2,6-diamine hydrochloride
CAS: 136-40-3; EC No: 205-243-8; MDL No: MFCD00035347; RTECS: US7875000
Crystalline; Molecular Formula: C11H11N5·HCl; MW: 249.70
Melting Point: 240° (decomposes)
- RTECS: US7875000
- Crystalline
Formula:
C11H12ClN5 MW: 249.70 g/mol Melting Pt: 240 °C (dec.) |
MDL Number:
MFCD00035347 CAS Number: 136-40-3 EINECS: 205-243-8 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...