2-Adamantanone oxime ≥97%
Supplier: BeanTown Chemical
140935-1G
140935-5G
BT140935-1GEA
106
USD
BT140935-1G
BT140935-5G
2-Adamantanone oxime ≥97%
2-Adamantanone oxime
CAS: 4500-12-3; EC No: 224-807-4; MDL No: MFCD00078273; RTECS: AU5018500
Solid; Molecular Formula: C10H15NO; MW: 165.23
Melting Point: 164-167°
Moisture Sensitive
- RTECS: AU5018500
- Moisture Sensitive
- Solid
Formula:
C₁₀H₁₅NO MW: 165.24 g/mol Melting Pt: 164…167 °C |
MDL Number:
MFCD00078273 CAS Number: 4500-12-3 EINECS: 224-807-4 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...