Methyl-3-nitrosalicylate ≥98%

Supplier: BeanTown Chemical

Synonyms: Methyl 2-hydroxy-3-nitrobenzoate

140925-1G 140925-25G 140925-5G
BT140925-1GEA 46 USD
BT140925-1G BT140925-25G BT140925-5G
Methyl-3-nitrosalicylate ≥98%
Methyl-3-nitrosalicylate

CAS: 22621-41-6; MDL No: MFCD00272263 Solid; Molecular Formula: C8H7NO5; MW: 197.15 Melting Point: 129-132°


  • Solid

Formula: C₈H₇NO₅
MW: 197.15 g/mol
Melting Pt: 129…132 °C
MDL Number: MFCD00272263
CAS Number: 22621-41-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR