Methyl-2-(triphenylphosphoranylidene)acetate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Methyl (triphenylphosphoranylidene)acetate, Methyl 2-(triphenylphosphoranylidene)acetate, Methoxycarbonylmethylene-Triphenylphosphorane
140890-100G
140890-25G
140890-500G
BT140890-100GEA
311.03
USD
BT140890-100G
BT140890-25G
BT140890-500G
Methyl-2-(triphenylphosphoranylidene)acetate ≥98%
Methyl-2-(triphenylphosphoranylidene)acetate
CAS: 2605-67-6; EC No: 220-018-4; MDL No: MFCD00008455
Crystalline; Linear Formula: (C6H5)3P=CHCO2CH3; Molecular Formula: C21H19O2P; MW: 334.35
Melting Point: 168-172°
Air Sensitive
- Air Sensitive
- Crystalline
Formula:
C₂₁H₁₉O₂P MW: 334.36 g/mol Melting Pt: 168…172 °C |
MDL Number:
MFCD00008455 CAS Number: 2605-67-6 EINECS: 220-018-4 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...