Ethyl-2-chloro-5-pyridylglyoxylate ≥97%

Supplier: BeanTown Chemical

140880-1G 140880-5G
BT140880-1GEA 205 USD
BT140880-1G BT140880-5G
Ethyl-2-chloro-5-pyridylglyoxylate ≥97%
Ethyl-2-chloro-5-pyridylglyoxylate

CAS: 902837-55-2; MDL No: MFCD07781188 Powder; Molecular Formula: C9H8ClNO3; MW: 213.62


  • Powder

Formula: C₉H₈ClNO₃
MDL Number: MFCD07781188
CAS Number: 902837-55-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR