Titanium(II) oxide ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Titanium oxide, Titanium monoxide

140855-100G 140855-25G 140855-5G
BT140855-100GEA 335 USD
BT140855-100G BT140855-25G BT140855-5G
Titanium(II) oxide ≥99.5%
Titanium(II) oxide

CAS: 12137-20-1; EC No: 235-236-5; MDL No: MFCD00036279 -325 Mesh Powder; Linear Formula: TiO; MW: 63.90 Melting Point: 1700°; Boiling Point: 3000° Density (g/mL): 4.95


  • -325 Mesh Powder

Formula: OTi
MW: 63.88
Boiling Pt: 3660
Melting Pt: 1750
Density: 4.95 g/mL
MDL Number: MFCD00036279
CAS Number: 12137-20-1
EINECS: 235-236-5

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR