N-Boc-4-oxo-L-proline methyl ester ≥97%
Supplier: BeanTown Chemical
CAS: 102195-80-2; MDL No: MFCD01861778
Powder; Molecular Formula: C11H17NO5; MW: 243.26
Melting Point: 42-46°; Flash point:
- Optical rotation: [α]22/D +14.0°, c = 1 in chloroform
- Air Sensitive, Heat Sensitive, Moisture Sensitive
- Powder
Formula:
C₁₁H₁₇NO₅ Melting Pt: 42…46 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD01861778 CAS Number: 102195-80-2 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...