Tris(ethylenediamine)rhodium(III) chloride trihydrate

Supplier: BeanTown Chemical

140840-1G 140840-500MG 140840-5G
BT140840-1GEA 401 USD
BT140840-1G BT140840-500MG BT140840-5G
Tris(ethylenediamine)rhodium(III) chloride trihydrate
Tris(ethylenediamine)rhodium(III) chloride trihydrate

CAS: 15004-86-1; EC No: 237-846-7; MDL No: MFCD00149661 Crystalline; Linear Formula: (NH2CH2CH2NH2)3RhCl3·3H2O; Molecular Formula: C6H24Cl3N6Rh·3H2O; MW: 443.61 Melting Point: 280° (decomposes)


  • Crystalline

Formula: C₆H₃₀Cl₃N₆O₃Rh
MW: 443.601 g/mol
Melting Pt: 280 °C (dec.)
MDL Number: MFCD00149661
CAS Number: 15004-86-1
EINECS: 237-846-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR