Benzyl bromoacetate ≥97%

Supplier: BeanTown Chemical

Synonyms: Benzyl 2-bromoacetate, Bromoacetic benzyl ester

140800-1KG 140800-250G 140800-50G
BT140800-1KGEA 332 USD
BT140800-1KG BT140800-250G BT140800-50G
Benzyl bromoacetate ≥97%
Benzyl bromoacetate

CAS: 5437-45-6; EC No: 226-611-4; MDL No: MFCD00000190; RTECS: AF5957215 Liquid; Linear Formula: C6H5CH2CO2CH2Br ; Molecular Formula: C9H9BrO2; MW: 229.07 Boiling Point: 166-170°/22 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.446; Refractive Index: 1.544


  • RTECS: AF5957215
  • Refractive index: 1.544
  • Liquid

Formula: C₉H₉BrO₂
MW: 229.07 g/mol
Boiling Pt: 166…170 °C/22 mmHg
Density: 1.446 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00000190
CAS Number: 5437-45-6
EINECS: 226-611-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR