2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose ≥98%

Supplier: BeanTown Chemical

Synonyms: D-Fructopyranose diacetonide, ((3aS,5aR,8aR,8bS)-2,2,7,7-Tetramethyltetrahydro-3aH-bis([1,3]dioxolo)[4,5-b:4',5'-d]pyran-3a-yl)methanol, Diacetonefructose

140785-10G 140785-50G
BT140785-10GEA 95 USD
BT140785-10G BT140785-50G
2,3:4,5-Di-O-isopropylidene-β-D-fructopyranose ≥98%
2,3:4,5-Di-O-isopropylidene-beta-D-fructopyranose

CAS: 20880-92-6; MDL No: MFCD00067639 Powder; Molecular Formula: C12H20O6; MW: 260.28


  • Powder

Formula: C₁₂H₂₀O₆
MW: 260.29 g/mol
MDL Number: MFCD00022183
CAS Number: 20880-92-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR