Ammonium sulfate ≥99.0% ACS

Supplier: BeanTown Chemical

Synonyms: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite

140780-100G 140780-2KG 140780-500G
BT140780-100GEA 26 USD
BT140780-100G BT140780-2KG BT140780-500G
Ammonium sulfate ≥99.0% ACS
Ammonium sulphate

CAS: 7783-20-2; EC No: 231-984-1; MDL No: MFCD00003391; RTECS: BS4500000 Granular; Linear Formula: (NH4)2SO4; MW: 132.14 Melting Point: 280° (decomposes) Density (g/mL): 1.77


  • RTECS: BS4500000
  • Granular

Formula: (NH₄)₂SO₄
MW: 132.14 g/mol
Melting Pt: 280 °C (dec.)
Density: 1.77 g/mL
MDL Number: MFCD00003391
CAS Number: 7783-20-2
EINECS: 231-984-1

Order Now

Specification Test Results

Assay 99.0% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR