trans-Dichlorobis(diethylsulfide)platinum(II)
Supplier: BeanTown Chemical
140760-1G
140760-5G
BT140760-1GEA
169
USD
BT140760-1G
BT140760-5G
trans-Dichlorobis(diethylsulfide)platinum(II)
trans-Dichlorobis(diethylsulfide)platinum(II)
CAS: 15337-84-5; EC No: 239-373-1; MDL No: MFCD00050943
Crystalline; Linear Formula: Pt[(CH2CH3)2S]2Cl2; Molecular Formula: C8H20Cl2PtS2; MW: 446.37
- Crystalline
Formula:
Pt[(C₂H₅)₂S]₂Cl₂ MW: 446.37 g/mol |
MDL Number:
MFCD00050943 CAS Number: 15337-84-5 EINECS: 239-373-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...