Methyl-3,3-dimethoxypropionate ≥96%
Supplier: BeanTown Chemical
Synonyms:
Methyl 3,3-dimethoxypropanoate, Methyl 3,3-dimethoxypropionate
140755-10G
140755-50G
BT140755-10GEA
35
USD
BT140755-10G
BT140755-50G
Methyl-3,3-dimethoxypropionate ≥96%
Methyl 3,3-dimethoxypropionate
CAS: 7424-91-1; EC No: 231-055-0; MDL No: MFCD00010650
Liquid; Linear Formula: (CH3O)2CHCH2COOCH3; Molecular Formula: C6H12O4; MW: 148.16
Boiling Point: 77°/20 mmHg; Flash point: 65°C (149°F)
Density (g/mL): 1.045; Refractive Index: 1.410
- Refractive index: 1.410
- Liquid
Formula:
C₆H₁₂O₄ MW: 148.16 g/mol Boiling Pt: 77 °C/20 mmHg Density: 1.045 g/mL Flash Pt: 65 °C (149 °F) |
MDL Number:
MFCD00010650 CAS Number: 7424-91-1 EINECS: 231-055-0 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...