Barium carbonate 99.0-101.0% ACS
Supplier: BeanTown Chemical
140750-1KG
140750-500G
BT140750-1KGEA
151.87
USD
BT140750-1KG
BT140750-500G
Barium carbonate 99.0-101.0% ACS
Barium carbonate
CAS: 513-77-9; EC No: 208-167-3; MDL No: MFCD00003448; RTECS: CQ8600000
Powder; Linear Formula: BaCO3; MW: 197.35
- RTECS: CQ8600000
- Powder
Formula:
BaCO₃ |
MDL Number:
MFCD00003448 CAS Number: 513-77-9 EINECS: 208-167-3 UN: 1564 ADR: 6.1,III |
Specification Test Results
Assay | 99.0-101.0% |
Insoluble in dilute hydrochloric acid | 0.015% min. |
Water soluble titratable base | ≤0.002meq/g |
Chloride (Cl) | ≤0.002% |
Oxidizing substance | ≤0.005% |
Sulfide (S) | ≤0.001% |
Calcium (Ca) | ≤0.05% |
Potassium (K) | ≤0.005% |
Sodium (Na) | ≤0.02% |
Strontium (Sr) | ≤0.7% |
Heavy metals | ≤0.001% |
Iron (Fe) | ≤0.002% |
Appearance | White powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...