Molybdenum ≥99.95% (trace metals basis), wire Ø 1.0 mm, annealed

Supplier: BeanTown Chemical

140725-100M 140725-25M 140725-5M
BT140725-100MEA 872 USD
BT140725-100M BT140725-25M BT140725-5M
Molybdenum ≥99.95% (trace metals basis), wire Ø 1.0 mm, annealed
Molybdenum

CAS: 7439-98-7; EC No: 231-107-2; MDL No: MFCD00003465; RTECS: QA4680000 Solid; Molecular Formula: Mo; MW: 95.94 Melting Point: 2617°; Boiling Point: 4612° Density (g/mL): 10.3


  • RTECS: QA4680000
  • Solid

Formula: Mo
MW: 95.94 g/mol
Boiling Pt: 3617
Melting Pt: 2622
Density: 10.3 g/mL
MDL Number: MFCD00003465
CAS Number: 7439-98-7
EINECS: 231-107-2
Merck Index: 12,06317

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR