Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II) ≥99%

Supplier: BeanTown Chemical

140720-10G 140720-2G 140720-500MG
BT140720-10GEA 1594 USD
BT140720-10G BT140720-2G BT140720-500MG
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II) ≥99%
Carbonyl(dihydrido)tris(triphenylphosphine)ruthenium(II)

CAS: 25360-32-1; MDL No: MFCD00015870 Crystalline; Linear Formula: Ru(CO)H2[P(C6H5)3]3; Molecular Formula: C55H47OP3Ru; MW: 919.79 Melting Point: 161-163° Light Sensitive, Moisture Sensitive


  • Light Sensitive, Moisture Sensitive
  • Crystalline

Formula: C₅₅H₄₇OP₃Ru
Melting Pt: 161…163 °C
MDL Number: MFCD00015870
CAS Number: 25360-32-1

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR