Magnesium aluminate ≥99.9% (trace metals basis excluding Ca)

Supplier: BeanTown Chemical

140680-100G 140680-25G 140680-5G
BT140680-100GEA 322 USD
BT140680-100G BT140680-25G BT140680-5G
Magnesium aluminate ≥99.9% (trace metals basis excluding Ca)

CAS: 12068-51-8; EC No: 235-100-5; MDL No: MFCD00049475 3-12mm Fused Pieces; Molecular Formula: Al2MgO4; MW: 142.26


  • 3-12mm Fused Pieces

Formula: Al₂MgO₄
MDL Number: MFCD00049475
CAS Number: 12068-51-8
EINECS: 235-100-5

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR