magnesium pyrophosphate

Supplier: BeanTown Chemical

140670-1KG 140670-250G 140670-50G
BT140670-1KGEA 454 USD
BT140670-1KG BT140670-250G BT140670-50G
magnesium pyrophosphate
magnesium pyrophosphate

CAS: 13446-24-7; EC No: 236-595-0; MDL No: MFCD00049482 Powder; Linear Formula: Mg2P2O7; MW: 222.57


  • Powder

Formula: Mg₂O₇P₂
MDL Number: MFCD00049482
CAS Number: 13446-24-7
EINECS: 236-595-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR