Bisacrylamide ≥99%

Supplier: BeanTown Chemical

Synonyms: Bis (N,N'-methylenebisacrylamide), N,N-Methylenebisacrylamide, N,N'-Methylenediacrylamide, N,N’-Methylenebisacrylamide, N,N'-Methylenebisacrylamide, BIS

140660-100G 140660-25G
BT140660-100GEA 110.41 USD
BT140660-100G BT140660-25G
Bisacrylamide ≥99%
Bisacrylamide

CAS: 110-26-9; EC No: 203-750-9; MDL No: MFCD00008625; RTECS: AS3678000 Powder; Linear Formula: (H2C=CHCONH)2CH2; Molecular Formula: C7H10N2O2; MW: 154.17 Melting Point: 300° Density (g/mL): 1.235 Air Sensitive, Light Sensitive


  • RTECS: AS3678000
  • Air Sensitive, Light Sensitive
  • Powder

Formula: (H₂C=CHCONH)₂CH₂
MW: 154.17 g/mol
Melting Pt: >300 °C
Density: 1.235 g/mL
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR