Graphite 99,5, foil, Thickness: 0.13 mm (0.005 in)

Supplier: BeanTown Chemical

140635-30X100 140635-30X30CM 140635-30X500
BT140635-30X10EA 126.59 USD
BT140635-30X10 BT140635-30X30 BT140635-30X50
Graphite 99,5, foil, Thickness: 0.13 mm (0.005 in)
Graphite

CAS: 7782-42-5; EC No: 231-955-3; MDL No: MFCD00144065 Solid; Molecular Formula: C; MW: 12.011


  • Solid

Formula: C
MW: 12.01 g/mol
Melting Pt: 3827
MDL Number: MFCD00144065
CAS Number: 7782-42-5
EINECS: 231-955-3
Merck Index: 12,04560

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR