5-Amino-2-bromopyridine ≥97%
Supplier: BeanTown Chemical
Synonyms:
6-Bromopyridin-3-amine, 2-Bromopyridin-5-amine, 3-Amino-6-bromopyridine, 2-Bromo-5-pyridylamine
140610-1G
140610-25G
140610-5G
BT140610-1GEA
42.57
USD
BT140610-1G
BT140610-25G
BT140610-5G
5-Amino-2-bromopyridine ≥97%
5-Amino-2-bromopyridine
CAS: 13534-97-9; EC No: 214-019-9; MDL No: MFCD01646116
Solid; Molecular Formula: C5H5BrN2 ; MW: 173.01
Melting Point: 74-76°
- Solid
Formula:
C₅H₅BrN₂ MW: 173.01 g/mol Melting Pt: 74…76 °C |
MDL Number:
MFCD01646116 CAS Number: 13534-97-9 EINECS: 214-019-9 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...