3-Chloro-4-iodopyridine ≥97%
Supplier: BeanTown Chemical
140600-1G
140600-5G
BT140600-1GEA
63
USD
BT140600-1G
BT140600-5G
3-Chloro-4-iodopyridine ≥97%
3-Chloro-4-iodopyridine
CAS: 77332-79-7; MDL No: MFCD00272199
Solid; Molecular Formula: C5H3ClIN; MW: 239.44
Melting Point: 104-108°
Light Sensitive
- Light Sensitive
- Solid
Formula:
C₅H₃ClIN MW: 239.44 g/mol Melting Pt: 104…108 °C |
MDL Number:
MFCD00272199 CAS Number: 77332-79-7 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...