(R)-(-)-Tetrahydrofurfuryl alcohol ≥98%

Supplier: BeanTown Chemical

Synonyms: (R)-Tetrahydro-2-furanmethanol, (R)-(Tetrahydrofuran-2-yl)methanol

140590-1G 140590-250MG
BT140590-1GEA 205 USD
BT140590-1G BT140590-250MG
(R)-(-)-Tetrahydrofurfuryl alcohol ≥98%
(R)-(-)-Tetrahydrofurfuryl alcohol

CAS: 22415-59-4; MDL No: MFCD03093085 Liquid; Molecular Formula: C5H10O2; MW: 102.13 Boiling Point: 178°; Flash point: 75°C (167°F) Density (g/mL): 1.054


  • Liquid

Formula: C₅H₁₀O₂
MW: 102.133 g/mol
Boiling Pt: 178 °C
Density: 1.054 g/mL
Flash Pt: 75 °C (167 °F)
MDL Number: MFCD03093085
CAS Number: 22415-59-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR