(4-Chlorophenyl)acetic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid

140575-1KG 140575-250G 140575-50G
BT140575-1KGEA 407 USD
BT140575-1KG BT140575-250G BT140575-50G
(4-Chlorophenyl)acetic acid ≥98%
(4-Chlorophenyl)acetic acid

CAS: 1878-66-6; EC No: 217-521-6; MDL No: MFCD01111378; RTECS: AG0590000 Powder; Molecular Formula: C8H7ClO2; MW: 170.59 Melting Point: 102-105°


  • RTECS: AG0590000
  • Powder

Formula: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Melting Pt: 102…105 °C
MDL Number: MFCD00004344
CAS Number: 1878-66-6
EINECS: 217-521-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR