Iron(III) phosphate hydrate
Supplier: BeanTown Chemical
Synonyms:
Iron phosphate hydrate, Iron (III) orthophosphate hydrate
140565-1KG
140565-250G
140565-5KG
BT140565-1KGEA
95
USD
BT140565-1KG
BT140565-250G
BT140565-5KG
Iron(III) phosphate hydrate
Iron(III) phosphate hydrate
CAS: 51833-68-2; EC No: 233-149-7; MDL No: MFCD00016096
Powder; Linear Formula: FePO4·xH2O; MW: 150.82 (anhydrous)
- Powder
Formula:
FeO₄P·xH₂O MW: 168.83 |
MDL Number:
MFCD00016096 CAS Number: 51833-68-2 EINECS: 233-149-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...