2-Cyano-3-methylpyridine ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-Methylpyridine-2-carbonitrile, 3-Methylpicolinonitrile, 2-Cyano-β-picoline, 2-Cyano-3-picoline
140545-100G
140545-25G
140545-5G
BT140545-100GEA
437
USD
BT140545-100G
BT140545-25G
BT140545-5G
2-Cyano-3-methylpyridine ≥98%
2-Cyano-3-methylpyridine
CAS: 20970-75-6; EC No: 244-131-3; MDL No: MFCD00190585
Solid; Molecular Formula: C7H6N2 ; MW: 118.14
Melting Point: 83-87°; Flash point: 110°C (230°F)
- Solid
Formula:
C₇H₆N₂ MW: 118.14 g/mol Melting Pt: 83…87 °C Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00190585 CAS Number: 20970-75-6 EINECS: 244-131-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...