Potassium stannate trihydrate ≥95%

Supplier: BeanTown Chemical

140520-1KG 140520-250G
BT140520-1KGEA 83 USD
BT140520-1KG BT140520-250G
Potassium stannate trihydrate ≥95%
Potassium stannate trihydrate

CAS: 12125-03-0; EC No: 234-721-9; MDL No: MFCD00150391; RTECS: TT5850000 Crystalline; Linear Formula: K2SnO3·3H2O; MW: 298.95 Melting Point: 140° (decomposes) Density (g/mL): 3.197


  • RTECS: TT5850000
  • Crystalline

Formula: K₂O₃Sn·3H₂O
Melting Pt: 140 °C (dec.)
Density: 3.197 g/mL
MDL Number: MFCD00150391
CAS Number: 12125-03-0
EINECS: 234-721-9

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR