5-Aminoindole ≥97%

Supplier: BeanTown Chemical

Synonyms: Indol-5-ylamine, 5-Indolamine, 5-Amino-1H-indole, 1H-Indol-5-amine

140510-1G 140510-25G 140510-5G
BT140510-1GEA 83 USD
BT140510-1G BT140510-25G BT140510-5G
5-Aminoindole ≥97%
5-Aminoindole

CAS: 5192-03-0; EC No: 225-977-2; MDL No: MFCD00005679 Crystalline; Molecular Formula: C8H8N2; MW: 132.16 Melting Point: 131-133° Air Sensitive, Light Sensitive


  • Air Sensitive, Light Sensitive
  • Crystalline

Formula: C₈H₈N₂
MW: 132.16 g/mol
Boiling Pt: 190
Melting Pt: 131…133 °C
MDL Number: MFCD00005679
CAS Number: 5192-03-0
EINECS: 225-977-2

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR