Carboxymethyl cellulose sodium salt

Supplier: BeanTown Chemical
Warning

140460-2.5KG 140460-500G
BT140460-2.5KGEA 231 USD
BT140460-2.5KG BT140460-500G
Carboxymethyl cellulose sodium salt
Carboxymethyl cellulose sodium salt

CAS: 9004-32-4; MDL No: MFCD00081472 Powder


  • Powder

Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number: MFCD00081472
CAS Number: 9004-32-4
Merck Index: 14,1829

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR