Phthalic acid ≥99.5% ACS
Supplier: BeanTown Chemical
Synonyms:
Pathalic acid
140430-100G
140430-500G
BT140430-100GEA
38.88
USD
BT140430-100G
BT140430-500G
Phthalic acid ≥99.5% ACS
Phthalic acid
CAS: 88-99-3; EC No: 201-873-2; MDL No: MFCD00002467; RTECS: TH9625000
Crystalline/Powder; Linear Formula: C6H4(COOH)2; Molecular Formula: C8H6O4; MW: 166.13
Melting Point: 201-211° (decomposes); Flash point: 168°C (334°F)
- RTECS: TH9625000
- Crystalline/Powder
Formula:
C₈H₆O₄ MW: 166.13 g/mol Melting Pt: 201…211 °C (dec.) Flash Pt: 168 °C (334 °F) |
MDL Number:
MFCD00002467 CAS Number: 88-99-3 EINECS: 201-873-2 |
Specification Test Results
Assay | 99.5% min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...