trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-Ethyl-4'-(trans-4-pentylcyclohexyl)biphenyl, 4-Ethyl-4'-(trans-4-pentylcyclohexyl)-1,1'-biphenyl
140410-1G
140410-5G
BT140410-1GEA
103
USD
BT140410-1G
BT140410-5G
trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl ≥97%
trans-4-Ethyl-4'-(4-n-pentylcyclohexyl)biphenyl
CAS: 79709-85-6; EC No: 279-240-5; MDL No: MFCD01941072
Powder; Molecular Formula: C25H34; MW: 334.54
Melting Point: 145-147°
- Powder
Formula:
C₂₅H₃₄ MW: 334.547 g/mol Melting Pt: 145…147 °C |
MDL Number:
MFCD01941072 CAS Number: 79709-85-6 EINECS: 279-240-5 |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...