3,3'-Diaminobenzidine (DAB) ≥99%

Supplier: BeanTown Chemical

Synonyms: [1,1'-Biphenyl]-3,3',4,4'-tetraamine, DAB, 3,3',4,4'-Biphenyltetramin, Biphenyl-3,3',4,4'-tetrayltetraamine

140400-100G 140400-25G 140400-5G
BT140400-100GEA 454 USD
BT140400-100G BT140400-25G BT140400-5G
3,3'-Diaminobenzidine (DAB) ≥99%
3,3'-Diaminobenzidine (DAB)

CAS: 91-95-2; EC No: 202-110-6; MDL No: MFCD00007725; RTECS: DV8750000 Solid; Molecular Formula: C12H14N4; MW: 214.27 Melting Point: 175-177° Light Sensitive


  • RTECS: DV8750000
  • Light Sensitive
  • Solid

Formula: C₁₂H₁₄N₄
Melting Pt: 175…177 °C
MDL Number: MFCD00007725
CAS Number: 91-95-2
EINECS: 202-110-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR