1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,1,1-Trichloro-2-methyl-2-propanol Hemihydrate, Chlorbutanol hemihydrate, Chlorobutanol hemihydrate, Chlorobutanol hydrous
140390-100G
140390-25G
BT140390-100GEA
89
USD
BT140390-100G
BT140390-25G
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate ≥98%
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate
CAS: 6001-64-5; EC No: 200-317-6; MDL No: MFCD00004461; RTECS: UC0175000
Solid; Molecular Formula: C4H7Cl3O·0.5H2O ; MW: 186.46
Melting Point: 77-79°; Flash point: 100°C (212°F)
- RTECS: UC0175000
- Solid
Formula:
C₈H₁₆Cl₆O₃ MW: 372.93 g/mol Melting Pt: 77…79 °C Flash Pt: 100 °C (212 °F) |
MDL Number:
MFCD02179352 CAS Number: 6001-64-5 EINECS: 200-317-6 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...