4-(Boc-amino)piperidine ≥96%
Supplier: BeanTown Chemical
Synonyms:
4-N-(tert-Butoxycarbonyl)aminopiperidine, tert-Butyl piperidin-4-ylcarbamate, tert-butyl N-(piperidin-4-yl)carbamate, 4-Boc-Aminopiperidine, 4-N-BOC-Aminopiperidine
140320-1G
140320-25G
140320-5G
BT140320-1GEA
48
USD
BT140320-1G
BT140320-25G
BT140320-5G
4-(Boc-amino)piperidine ≥96%
4-(Boc-amino)piperidine
CAS: 73874-95-0; MDL No: MFCD00798171
Solid; Molecular Formula: C10H20N2O2; MW: 200.28
Melting Point: 162-166°
Air Sensitive
- Air Sensitive
- Solid
Formula:
C₁₀H₂₀N₂O₂ MW: 200.28 g/mol Melting Pt: 162…166 °C |
MDL Number:
MFCD00798171 CAS Number: 73874-95-0 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...