4-Bromo-3-methylanisole ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Bromo-4-methoxy-2-methylbenzene, 2-Bromo-5-methoxytoluene

140295-10G 140295-250G 140295-50G
BT140295-10GEA 53 USD
BT140295-10G BT140295-250G BT140295-50G
4-Bromo-3-methylanisole ≥97%
4-Bromo-3-methylanisole

CAS: 27060-75-9; MDL No: MFCD00060687 Liquid; Molecular Formula: C8H9BrO; MW: 201.06 Boiling Point: 111-112°/15 mmHg; Flash point: 110°C (230°F) Density (g/mL): 1.424; Refractive Index: 1.561


  • Refractive index: 1.561
  • Liquid

Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 111…112 °C/15 mmHg
Density: 1.424 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00060687
CAS Number: 27060-75-9

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR