2-Cyanobenzoic acid ≥96%
Supplier: BeanTown Chemical
140275-1G
140275-5G
BT140275-1GEA
37
USD
BT140275-1G
BT140275-5G
2-Cyanobenzoic acid ≥96%
2-Cyanobenzoic acid
CAS: 3839-22-3; MDL No: MFCD00045796
Powder; Molecular Formula: C8H5NO2; MW: 147.13
Melting Point: 212°
- Powder
Formula:
C₈H₅NO₂ Melting Pt: 212 °C |
MDL Number:
MFCD00045796 CAS Number: 3839-22-3 |
Specification Test Results
Assay | 96% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...