3-Hydroxypyrazine-2-carboxamide ≥98%

Supplier: BeanTown Chemical

Synonyms: T-1105

140260-1G 140260-250MG 140260-5G
BT140260-1GEA 100 USD
BT140260-1G BT140260-250MG BT140260-5G
3-Hydroxypyrazine-2-carboxamide ≥98%
3-Hydroxypyrazine-2-carboxamide

CAS: 55321-99-8; MDL No: MFCD00233977 Crystalline/Powder; Molecular Formula: C5H5N3O2; MW: 139.12 Melting Point: 270° (decomposes)


  • Crystalline/Powder

Formula: C₅H₅N₃O₂
MW: 139.114 g/mol
Melting Pt: 270 °C (dec.)
MDL Number: MFCD00233977
CAS Number: 55321-99-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR