5-Bromopyrimidine ≥98%

Supplier: BeanTown Chemical

140255-100G 140255-25G 140255-5G
BT140255-100GEA 207 USD
BT140255-100G BT140255-25G BT140255-5G
5-Bromopyrimidine ≥98%
5-Bromopyrimidine

CAS: 4595-59-9; EC No: 224-992-1; MDL No: MFCD00006117 Solid; Molecular Formula: C4H3BrN2; MW: 158.98 Melting Point: 67-73°


  • Solid

Formula: C₄H₃BrN₂
MW: 158.99 g/mol
Melting Pt: 67…73 °C
MDL Number: MFCD00006117
CAS Number: 4595-59-9
EINECS: 224-992-1

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR