Calcium chloride, anhydrous ≥96.0% ACS

Supplier: BeanTown Chemical

140230-250G 140230-50G
BT140230-250GEA 88 USD
BT140230-250G BT140230-50G
Calcium chloride, anhydrous ≥96.0% ACS
Calcium chloride
Formula: CaCl₂
Melting Pt: 772 °C
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8

Order Now

Specification Test Results

Assay 96.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR