2,2,3,3,4,4,5,5-Octafluoro-1-pentanol ≥98%

Supplier: BeanTown Chemical

Synonyms: 1H,1H,5H-Octafluoro-1-pentanol

140195-100G 140195-25G 140195-500G
BT140195-100GEA 60 USD
BT140195-100G BT140195-25G BT140195-500G
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol ≥98%
2,2,3,3,4,4,5,5-Octafluoro-1-pentanol

CAS: 355-80-6; EC No: 206-593-4; MDL No: MFCD00039631; RTECS: SA7650000 Liquid; Linear Formula: F2CH(CF2)3CH2OH; Molecular Formula: C5H4F8O; MW: 232.07 Boiling Point: 141-142°; Flash point: 76°C (169°F) Density (g/mL): 1.667; Refractive Index: 1.318


  • RTECS: SA7650000
  • Refractive index: 1.318
  • Liquid

Formula: C₅H₄F₈O
MW: 232.07 g/mol
Boiling Pt: 141…142 °C
Density: 1.667 g/mL
Flash Pt: 76 °C (169 °F)
MDL Number: MFCD00039631
CAS Number: 355-80-6
EINECS: 206-593-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR