D-(+)-Malic acid ≥98%
Supplier: BeanTown Chemical
140180-1G
140180-25G
140180-5G
BT140180-1GEA
31
USD
BT140180-1G
BT140180-25G
BT140180-5G
D-(+)-Malic acid ≥98%
D-(+)-Malic acid
CAS: 636-61-3; EC No: 211-262-2; MDL No: MFCD00004245; RTECS: ON7260000
Powder; Molecular Formula: C4H6O5; MW: 134.09
Melting Point: 98-102°
Optical Rotation: [α]20/D +28.0±2°, c = 5.5% in pyridine
- RTECS: ON7260000
- Optical rotation: [α]20/D +28.0±2°, c = 5.5% in pyridine
- Powder
Formula:
C₄H₆O₅ Melting Pt: 98…102 °C |
MDL Number:
MFCD00004245 CAS Number: 636-61-3 EINECS: 211-262-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...